N-Boc-L-alaninol ≥98.0% (by GC, total nitrogen)
Fornitore: TCI EUROPE
Synonyms:
Boc-L-Alaninol, Boc-Alaninol, (S)-2-(BOC-amino)-1-propanol
B1953-1G
B1953-5G
TCIAB1953-1GEA
18
EUR
TCIAB1953-1G
TCIAB1953-5G
N-Boc-L-alaninol ≥98.0% (by GC, total nitrogen)
N-Boc-L-alaninol
Formula:
C₈H₁₇NO₃ Peso molecolare: 175,2 g/mol |
MDL Number:
MFCD00043121 CAS Number: 79069-13-9 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 57.0 to 61.0 deg-C |
Specific rotation [a]20/D | -8.1 to -9.1 deg (C=1.2, MeOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...