Triglycidyl isocyanurate ≥98.0% (by GC, titration analysis)
Fornitore: TCI
|
Pericolo
|
I0428-25G
I0428-500G
I0428-100G
TCIAI0428-25GEA
0
EUR
TCIAI0428-25G
TCIAI0428-500G
TCIAI0428-100G
Triglycidyl isocyanurate ≥98.0% (by GC, titration analysis)
Triglycidyl isocyanurate
Formula:
C₁₂H₁₅N₃O₆ |
MDL Number:
MFCD00080670 CAS Number: 2451-62-9 EINECS: 219-514-3 UN: 2811 ADR: 6.1,III |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Epoxy Equivalent) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...