Stampa…
Bathophenanthrolinedisulphonic acid disodium salt trihydrate, Supelco®
Fornitore: Merck
M-Clarity™ quality level = MQ100
11890-1G 11890-5G
11890-1GEA 280 EUR
11890-1G 11890-5G
Bathophenanthrolinedisulphonic acid disodium salt trihydrate, Supelco®
Bathophenanthrolinedisulphonic acid disodium salt trihydrate
Formula: C₂₄H₁₄N₂Na₂O₆S₂·3H₂O
MDL Number: MFCD00004977
CAS Number: 53744-42-6 (anhy)
EINECS: 258-152-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=143589d69ba19eb7217f4d6039ad34cfeaae9200&vid=fb64afbf91a456a657647d1e3efb1477ca8561a7&prid=web&clid=SAPDAM