Stampa…

5-(Trifluoromethoxy)isatin ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: 5-(Trifluoromethoxy)indoline-2,3-dione

T2774-1G T2774-5G
TCIAT2774-1GEA 31 EUR
TCIAT2774-1G TCIAT2774-5G
5-(Trifluoromethoxy)isatin ≥98.0% (by GC, titration analysis)
5-(Trifluoromethoxy)isatin
Formula: C₉H₄F₃NO₃
Peso molecolare: 231,13 g/mol
MDL Number: MFCD00192524
CAS Number: 169037-23-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=15df7f66574583898e81a5d02983a949508f75bb&vid=15df7f66574583898e81a5d02983a949508f75bb&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 169.0 to 173.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR