α-Methylcinnamic acid ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2-Methyl-3-phenyl-2-propenoic acid
M1336-25G
M1336-5G
TCIAM1336-25GEA
53
EUR
TCIAM1336-25G
TCIAM1336-5G
α-Methylcinnamic acid ≥98.0% (by GC, titration analysis)
α-Methylcinnamic acid
Formula:
C₁₀H₁₀O₂ Peso molecolare: 162,19 g/mol Punto di ebollizione: 288°C Punto di fusione: 80°C |
MDL Number:
MFCD00002652 CAS Number: 1199-77-5 EINECS: 214-847-0 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 79.0 to 83.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...