Stampa…

Benzyltrimethylammonium tribromide ≥97.0% (by titrimetric analysis) brominating reagent

Fornitore: TCI
Attenzione

Synonyms: Mono(N,N,N-trimethyl-1-phenylmethanaminium) tribromide, Benzyl trimethyl ammonium tribromide

T1382-25G T1382-5G
TCIAT1382-25GEA 51 EUR
TCIAT1382-25G TCIAT1382-5G
Benzyltrimethylammonium tribromide ≥97.0% (by titrimetric analysis) brominating reagent
Benzyltrimethylammonium tribromide
Formula: C₁₀H₁₆Br₃N
Peso molecolare: 389.97 g/mol
MDL Number: MFCD00134423
CAS Number: 111865-47-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d688bac2ea9376eb310debf2cdc9dfda68926581&vid=88a2356b08a12a6cf30d5b138c552d49dcd9516e&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown powder to crystal
Purity (Iodometric Titration) min. 97.0 %
Melting point 100.0 to 103.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR