(S)-(+)-2-(Methoxymethyl)pyrrolidine ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
(S)-2-(Methoxymethyl)pyrrolidine
M1161-1G
M1161-5G
TCIAM1161-1GEA
34
EUR
TCIAM1161-1G
TCIAM1161-5G
(S)-(+)-2-(Methoxymethyl)pyrrolidine ≥98.0% (by GC)
(S)-(+)-2-(Methoxymethyl)pyrrolidine
Formula:
C₆H₁₃NO Peso molecolare: 115,18 g/mol Punto di ebollizione: 68°C Flash Pt: 45°C |
MDL Number:
MFCD00010408 CAS Number: 63126-47-6 UN: 1993 ADR: 3,III |
Risultati del test di specificazione
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Specific gravity (20/20) | 0.9320 to 0.9380 |
Refractive index n20/D | 1.4450 to 1.4470 |
Specific rotation [a]20/D | +35.0 to +41.0 deg (C=2, 1mol/L HCl) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...