(R)-(-)-2-(Methoxymethyl)pyrrolidine ≥99.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
(R)-2-(Methoxymethyl)pyrrolidine
M1169-1G
M1169-5G
TCIAM1169-1GEA
89
EUR
TCIAM1169-1G
TCIAM1169-5G
(R)-(-)-2-(Methoxymethyl)pyrrolidine ≥99.0% (by GC)
(R)-(-)-2-(Methoxymethyl)pyrrolidine
Formula:
C₆H₁₃NO Peso molecolare: 115,18 g/mol Flash Pt: 45°C |
MDL Number:
MFCD00066219 CAS Number: 84025-81-0 UN: 1993 ADR: 3,III |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 99.0 % |
Specific rotation [a]20/D | -33.0 to -38.0 deg (C=2, 1mol/L HCl) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...