(S)-(+)-3-Aminopyrrolidine dihydrochloride ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
(S)-Pyrrolidin-3-amine dihydrochloride
A1054-25G
A1054-5G
TCIAA1054-25GEA
437
EUR
TCIAA1054-25G
TCIAA1054-5G
(S)-(+)-3-Aminopyrrolidine dihydrochloride ≥98.0% (by titrimetric analysis)
(S)-(+)-3-Aminopyrrolidine dihydrochloride
Formula:
C₄H₁₂Cl₂N₂ Peso molecolare: 159,06 g/mol |
MDL Number:
MFCD00070605 CAS Number: 116183-83-6 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (Argentmetric Titration) | min. 98.0 % |
Specific rotation [a]20/D | +0.9 to +1.3 deg (C=10, H2O) |
Solubility in Water | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...