Stampa…

Bathocuproin ≥99.0% (by HPLC, titration analysis), purified by sublimation

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2,9-Dimethyl-4,7-diphenyl-1,10-phenanthroline

B2694-1G B2694-5G
TCIAB2694-1GEA 120 EUR
TCIAB2694-1G TCIAB2694-5G
Bathocuproin ≥99.0% (by HPLC, titration analysis), purified by sublimation
Bathocuproin
Formula: C₂₆H₂₀N₂
Peso molecolare: 360,46 g/mol
Punto di fusione: 283°C
MDL Number: MFCD00004972
CAS Number: 4733-39-5
EINECS: 225-240-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=17474c845dab44cce4189a350cd9237533d79406&vid=9a24e98d5e8fc38cb93a10574e300b2b790a3cb3&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Green powder to crystal
Purity (HPLC) min. 99.0 area%
Purity (Nonaqueous Titration) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR