Methyl-4-(chloromethyl)benzoate ≥98.0% (by GC)
Fornitore: TCI
|
Pericolo
|
Synonyms:
Methyl 4-(chloromethyl)benzoate
Formula:
C₉H₉ClO₂ Peso molecolare: 184.62 g/mol Punto di ebollizione: 165°C Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00511224 CAS Number: 34040-64-7 EINECS: 251-806-6 UN: 3261 ADR: 8 III |
|
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 37.0 to 41.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...