6-Bromopicolinamide ≥97%
Fornitore: Thermo Fisher Scientific
|
Attenzione
|
Synonyms:
6-Bromopyridine-2-carboxamide
Purity: 97%
CAS No.: 25194-52-9
Molecular Formula: C6H5BrN2O
Formula Weight: 201.02
MDL No.: MFCD00234154
Formula:
C₆H₅BrN₂O Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00234154 CAS Number: 25194-52-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...