L-Methionine sulphone ≥99%
Fornitore: Thermo Scientific
Synonyms:
(S)-2-Amino-4-(methylsulphonyl)butanoic acid
A17027.03
A17027.06
A17027.03EA
47
EUR
A17027.03
A17027.06
L-Methionine sulphone ≥99%
L-Methionine sulphone
Purity: 99+%
CAS No.: 7314-32-1
Molecular Formula: C5H11NO4S
Formula Weight: 181.21
Melting Point: 254° dec.
Beilstein Reference: 1725510
Formula:
C₅H₁₁NO₄S Peso molecolare: 181,21 g/mol Punto di fusione: 254 °C (dec.) Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00066020 CAS Number: 7314-32-1 EINECS: 230-774-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...