Stampa…

N-Formyl-DL-ethionine ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

F0120-1G F0120-100MG
TCIAF0120-1GEA 324 EUR
TCIAF0120-1G TCIAF0120-100MG
N-Formyl-DL-ethionine ≥98.0% (by HPLC, titration analysis)
N-Formyl-DL-ethionine
Formula: C₇H₁₃NO₃S
MDL Number: MFCD00059815
CAS Number: 126872-00-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ffc87ab95a1b3c1a1d4b5c42bfc413973c215cc8&vid=ffc87ab95a1b3c1a1d4b5c42bfc413973c215cc8&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point gt. 115.0 to 119.0 deg-C
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR