Stampa…

(±)-1-Boc-3-aminopyrrolidine ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 3-Aminopyrrolidine, N1-BOC protected, 1-Boc-3-Aminopyrrolidine

B3890-5G B3890-1G
TCIAB3890-5GEA 294 EUR
TCIAB3890-5G TCIAB3890-1G
(±)-1-Boc-3-aminopyrrolidine ≥98.0% (by GC, titration analysis)
(±)-1-Boc-3-aminopyrrolidine
Formula: C₉H₁₈N₂O₂
Peso molecolare: 186,25 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD01861220
CAS Number: 186550-13-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=50ed788098112ee46708bd685bd75d0af0b6bbf5&vid=50ed788098112ee46708bd685bd75d0af0b6bbf5&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR