Stampa…

3-Amino-1-Boc-piperidine ≥97.0% (by GC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: 1-BOC-3-Aminopiperidine, (±)-3-Amino-1-boc-piperidine, tert-Butyl 3-aminopiperidine-1-carboxylate

A2314-5G A2314-1G
TCIAA2314-5GEA 112 EUR
TCIAA2314-5G TCIAA2314-1G
3-Amino-1-Boc-piperidine ≥97.0% (by GC, titration analysis)
3-Amino-1-Boc-piperidine
Formula: C₁₀H₂₀N₂O₂
Peso molecolare: 200,28 g/mol
MDL Number: MFCD01861219
CAS Number: 184637-48-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e69ba73997d4a075cb7556979f4d14a29b12c69c&vid=e69ba73997d4a075cb7556979f4d14a29b12c69c&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR