Stampa…
HPLC columns, Hypersil™ Phenyl and Phenyl-2
Fornitore: Thermo Fisher Scientific
31905-254630 30905-254630 30905-254030 30905-154630 30905-014001 30903-154630 30903-153030 30903-014001
555-5410EA 841 EUR
555-5410 555-5304 555-5305 555-5308 555-6421 555-6424 555-6426 555-6709
HPLC columns, Hypersil™ Phenyl and Phenyl-2
Colonne cromatografiche
These columns are used to analyse the aromatic and moderately polar compounds.
  • Hypersil Phenyl-2 is endcapped
  • Alternative selectivity to C8
Visualizza i codici
Specifiche
Environmentally Preferable
Carico di carbonio 5% 5% 5%
Con endcapping No No
Pressione max. 400 bar 400 bar 400 bar
Range di pH 2 - 8 2 - 8 2 - 8
Area superficiale 170 m²/g 170 m²/g 170 m²/g
Limite di temperatura 60 °C 60 °C 60 °C
Codice USP L11 L11