Stampa…
Workers jackets, C.I., Design C, true navy
Fornitore: FRISTADS KANSAS
8495391312XL 849539131L 849539131M 849539131S 849539131XL 849539131XS 8495392102XL 849539210L 849539210M 849539210S 849539210XL 849539210XS 8495392312XL 849539231L 849539231M 849539231S 849539231XL 849539231XS 8495393312XL 849539331L 849539331M 849539331S 849539331XL 849539331XS 8495394402XL 849539440L 849539440M 849539440S 849539440XL 849539440XS 8495395202XL 849539520L 849539520M 849539520S 849539520XL 849539520XS 8495395302XL 849539530L 849539530M 849539530S 849539530XL 849539530XS 8495395392XL 849539539L 849539539M 849539539S 849539539XL 849539539XS 8495396402XL 849539640L 849539640M 849539640S 849539640XL 849539640XS 8495397302XL 849539730L 849539730M 849539730S 849539730XL 849539730XS 8495399402XL 849539940L 849539940M 849539940S 849539940XL 849539940XS 8495399412XL 849539941L 849539941M 849539941S 849539941XL 849539941XS
KWIN8495391312XLEA 0 EUR
KWIN8495391312XL KWIN849539131L KWIN849539131M KWIN849539131S KWIN849539131XL KWIN849539131XS KWIN8495392102XL KWIN849539210L KWIN849539210M KWIN849539210S KWIN849539210XL KWIN849539210XS KWIN8495392312XL KWIN849539231L KWIN849539231M KWIN849539231S KWIN849539231XL KWIN849539231XS KWIN8495393312XL KWIN849539331L KWIN849539331M KWIN849539331S KWIN849539331XL KWIN849539331XS KWIN8495394402XL KWIN849539440L KWIN849539440M KWIN849539440S KWIN849539440XL KWIN849539440XS KWIN8495395202XL KWIN849539520L KWIN849539520M KWIN849539520S KWIN849539520XL KWIN849539520XS KWIN8495395302XL KWIN849539530L KWIN849539530M KWIN849539530S KWIN849539530XL KWIN849539530XS KWIN8495395392XL KWIN849539539L KWIN849539539M KWIN849539539S KWIN849539539XL KWIN849539539XS KWIN8495396402XL KWIN849539640L KWIN849539640M KWIN849539640S KWIN849539640XL KWIN849539640XS KWIN8495397302XL KWIN849539730L KWIN849539730M KWIN849539730S KWIN849539730XL KWIN849539730XS KWIN8495399402XL KWIN849539940L KWIN849539940M KWIN849539940S KWIN849539940XL KWIN849539940XS KWIN8495399412XL KWIN849539941L KWIN849539941M KWIN849539941S KWIN849539941XL KWIN849539941XS
Workers jackets, C.I., Design C, true navy
Abbigliamento da lavoro
Industrial jacket made of 65% polyester and 35% cotton, with soft nap on reverse side. The navy blue jacket is available with different accent and piping colours.
  • Zip closure
  • Two chest pockets, one double with flap and velcro closure
  • Two front pockets, glove pocket, sleeve pocket, concealed ID-card pocket
  • Mobile phone pocket with loop for earpiece, concealed pen pocket, two inside pockets
  • Adjustable waist and cuffs, rounded elbows
  • Choice of 12 profille colours and 13 piping colours
Weight: 260 g/m²