Stampa…

Propagyl bromide 80% ∼9.2 mol/L in toluene stabilizzato

Fornitore: TCI EUROPE
Pericolo

Synonyms: 3-Bromopropyne

Stabilization: Stabiised with magnesium oxide (MgO)

P1272-25G P1272-500G P1272-100G
TCIAP1272-25GEA 26 EUR
TCIAP1272-25G TCIAP1272-500G TCIAP1272-100G
Propagyl bromide 80% ∼9.2 mol/L in toluene stabilizzato
Propagyl bromide
Formula: C₃H₃Br
Peso molecolare: 118,96 g/mol
Flash Pt: 18°C
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00000241
CAS Number: 106-96-7
EINECS: 203-447-1
UN: 1993
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=250033596f589699867a5c38ba00e1349a4ab28a&vid=0d60dabac59081ffd643f22778ef600200ef8d11&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light orange to Yellow clear liquid
Concentration (GC) 80.0 to 83.0 w/w%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR