3-Bromophthalide ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
B1068-25G
B1068-5G
TCIAB1068-25GEA
136
EUR
TCIAB1068-25G
TCIAB1068-5G
3-Bromophthalide ≥98.0% (by titrimetric analysis)
3-Bromophthalide
Formula:
C₈H₅BrO₂ Peso molecolare: 213,0281 g/mol Punto di ebollizione: 138°C |
MDL Number:
MFCD00040520 CAS Number: 6940-49-4 EINECS: 230-084-6 |
Risultati del test di specificazione
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 95.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Melting point | 83.0 to 87.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...