5-Bromophthalide ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
5-Bromoisobenzofuran-1(3H)-one
B2770-25G
B2770-5G
TCIAB2770-25GEA
109
EUR
TCIAB2770-25G
TCIAB2770-5G
5-Bromophthalide ≥98.0% (by GC)
5-Bromophthalide
Formula:
C₈H₅BrO₂ Peso molecolare: 213,03 g/mol |
MDL Number:
MFCD01797360 CAS Number: 64169-34-2 EINECS: 447-190-5 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 164.0 to 168.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...