Stampa…

Methyl-4-chloropicolinate ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Methyl 4-chloropicolinate, Methyl 4-chloropyridine-2-carboxylate

M1832-1G M1832-5G
TCIAM1832-1GEA 30 EUR
TCIAM1832-1G TCIAM1832-5G
Methyl-4-chloropicolinate ≥98.0% (by GC, titration analysis)
Methyl-4-chloropicolinate
Formula: C₇H₆ClNO₂
Peso molecolare: 171,5800 g/mol
MDL Number: MFCD04116183
CAS Number: 24484-93-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bb887dc521dd803d391773267a3f4dd6ac2ac072&vid=bb887dc521dd803d391773267a3f4dd6ac2ac072&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 57.0 to 61.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR