2-Chloro-2-methylnonane ≥90%
Fornitore: Thermo Scientific
|
Attenzione
|
43784.14
43784.06
43784.14EA
282
EUR
43784.14
43784.06
2-Chloro-2-methylnonane ≥90%
2-Chloro-2-methylnonane
Purity: 90+%
CAS No.: 4325-50-2
Molecular Formula: CH3(CH2)6C(CH3)(Cl)CH3
Formula Weight: 176.73
Physical Form: Liquid
Appearance: Colourless
UN No.: 3082
MDL No.: MFCD00060781
Formula:
C₁₀H₂₁Cl Peso molecolare: 176,73 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00060781 CAS Number: 4325-50-2 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...