Stampa…

4,4'-Diacetylbiphenyl ≥99.0% (by GC)

Fornitore: TCI
Attenzione

D1902-10G D1902-25G
TCIAD1902-10GEA 60 EUR
TCIAD1902-10G TCIAD1902-25G
4,4'-Diacetylbiphenyl ≥99.0% (by GC)
4,4'-Diacetylbiphenyl
Formula: C₁₆H₁₄O₂
MDL Number: MFCD00017248
CAS Number: 787-69-9
UN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=74352fcbab30adbbe4a628ba173761b5b8fa466a&vid=74352fcbab30adbbe4a628ba173761b5b8fa466a&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 99.0 %
Melting point 192.0 to 195.0 deg-C
Solubility in Chloroform within almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR