Stampa…

4-Bromo-4'-propylbiphenyl ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 4-Bromo-4'-propyl-1,1'-biphenyl

B1306-25G B1306-5G
TCIAB1306-25GEA 84 EUR
TCIAB1306-25G TCIAB1306-5G
4-Bromo-4'-propylbiphenyl ≥98.0% (by GC)
4-Bromo-4'-propylbiphenyl
Formula: C₁₅H₁₅Br
Peso molecolare: 275,1800 g/mol
MDL Number: MFCD00060105
CAS Number: 58743-81-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=325b1bfb74c55f2d0b437adfe3932c5760912ee8&vid=325b1bfb74c55f2d0b437adfe3932c5760912ee8&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Solubility in Toluene almost transparency
Melting point 107.0 to 109.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR