Stampa…

4-Cyano-4'-heptylbiphenyl ≥95.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-Cyano-4'-n-heptylbiphenyl, 4'-Heptyl-[1,1'-biphenyl]-4-carbonitrile

H0812-1G H0812-5G
TCIAH0812-1GEA 32 EUR
TCIAH0812-1G TCIAH0812-5G
4-Cyano-4'-heptylbiphenyl ≥95.0% (by GC)
4-Cyano-4'-heptylbiphenyl
Formula: C₂₀H₂₃N
Peso molecolare: 277,41 g/mol
MDL Number: MFCD00074915
CAS Number: 41122-71-8
EINECS: 255-229-0

Visualizza i codici

Risultati del test di specificazione

Appearance White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
Purity (GC) min. 95.0 %
Mesomorphic range 27.0 to 45.0 deg-C
Solubility in Toluene almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR