Stampa…

Ethyl thiophene-2-carboxylate ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: Ethyl 2-thiophenecarboxylate

T0904-10ML T0904-25ML
TCIAT0904-10MLEA 24 EUR
TCIAT0904-10ML TCIAT0904-25ML
Ethyl thiophene-2-carboxylate ≥98.0% (by GC)
Ethyl thiophene-2-carboxylate
Formula: C₇H₈O₂S
Peso molecolare: 156,20 g/mol
Punto di ebollizione: 218°C
Flash Pt: 88°C
MDL Number: MFCD00005436
CAS Number: 2810-04-0
EINECS: 220-554-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ff1c4df905ecaf62ad71d4941455351073101ed3&vid=ff1c4df905ecaf62ad71d4941455351073101ed3&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR