Stampa…

2-Methoxy-6-methylpyridine ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

M1695-25G M1695-5G
TCIAM1695-25GEA 68 EUR
TCIAM1695-25G TCIAM1695-5G
2-Methoxy-6-methylpyridine ≥98.0% (by GC, titration analysis)
2-Methoxy-6-methylpyridine
Formula: C₇H₉NO
Peso molecolare: 123,16 g/mol
Punto di ebollizione: 156°C
MDL Number: MFCD04112507
CAS Number: 63071-03-4
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f910830deded617fd764c77a050926875ecf81b1&vid=f910830deded617fd764c77a050926875ecf81b1&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR