1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI
Formula:
C₂₀H₁₃O₄P |
MDL Number:
MFCD00010045 CAS Number: 35193-63-6 EINECS: 252-425-8 |
|
Risultati del test di specificazione
Appearance | White to Yellow to Orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in N,N-DMF | within almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...