Stampa…
(S)-(+)-1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate ≥98%
Fornitore: Thermo Scientific
Attenzione
Synonyms: (S)-(+)-1,1'-Binaphthalene-2,2'-diyl-phosphate, (11bS)-4-Hydroxydinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide
L14137.03 L14137.06
L14137.03EA 70 EUR
L14137.03 L14137.06
(S)-(+)-1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate ≥98%
(S)-(+)-1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate
Purity: 98+% CAS No.: 35193-64-7 Molecular Formula: C20H13O4P Formula Weight: 348.30 Melting Point: 300° Beilstein Reference: 4787775
Formula: C₂₀H₁₃O₄P
Peso molecolare: 348,289 g/mol
Punto di fusione: >300 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00010045
CAS Number: 35193-64-7