(R)-(-)-1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate ≥98.0% (by titrimetric analysis)
Fornitore: TCI
Formula:
C₂₀H₁₃O₄P |
MDL Number:
MFCD00010045 CAS Number: 39648-67-4 |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Optical purity (LC) | min. 98.0 ee% |
Purity (Neutralization titration) | min. 98.0 % |
Specific rotation [a]20/D | -590.0 to -610.0 deg (C=1, MeOH) |
Solubility in hot Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...