Stampa…

4-Cyano-4'-pentyloxybiphenyl ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-Amyloxy-4'-cyanobiphenyl, 4-Cyano-4'-n-pentyloxybiphenyl, 5OCB, 4'-Amyloxy-4-biphenylcarbonitrile, 4'-Pentyloxy-4-biphenylcarbonitrile, 4'-Pentyloxy-[1,1'-biphenyl]-4-carbonitrile

C1551-1G C1551-5G
TCIAC1551-1GEA 32 EUR
TCIAC1551-1G TCIAC1551-5G
4-Cyano-4'-pentyloxybiphenyl ≥98.0% (by GC)
4-Cyano-4'-pentyloxybiphenyl
Formula: C₁₈H₁₉NO
Peso molecolare: 265,35 g/mol
MDL Number: MFCD00074879
CAS Number: 52364-71-3
EINECS: 257-875-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c0e9a703e69c1817a4f4f5fc4d2daf08e4c7f66d&vid=c0e9a703e69c1817a4f4f5fc4d2daf08e4c7f66d&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystaline
Purity (GC) min. 98.0 %
Mesomorphic range 47.0 to 71.0 deg-C
Solubility in Toluene very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR