Stampa…

4-(Aminomethyl)benzonitrile hydrochloride ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-(Aminomethyl)benzonitrile HCl

A1645-1G A1645-5G
TCIAA1645-1GEA 17 EUR
TCIAA1645-1G TCIAA1645-5G
4-(Aminomethyl)benzonitrile hydrochloride ≥98.0% (by HPLC, titration analysis)
4-(Aminomethyl)benzonitrile hydrochloride
Formula: C₈H₉ClN₂
Peso molecolare: 168,62 g/mol
MDL Number: MFCD01861472
CAS Number: 15996-76-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5aa5c1093148befc42ec603f5c3735fc28d8471c&vid=5aa5c1093148befc42ec603f5c3735fc28d8471c&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Argentmetric Titration) min. 98.0 %
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR