Stampa…

(Dimethylamino)acetonitrile ≥96.0% (by GC, titration analysis)

Fornitore: TCI
Pericolo

D1929-25ML D1929-500ML
TCIAD1929-25MLEA 50 EUR
TCIAD1929-25ML TCIAD1929-500ML
(Dimethylamino)acetonitrile ≥96.0% (by GC, titration analysis)
(Dimethylamino)acetonitrile
Formula: C₄H₈N₂
Peso molecolare: 84.12 g/mol
Punto di ebollizione: 138°C
Flash Pt: 36°C
MDL Number: MFCD00001890
CAS Number: 926-64-7
EINECS: 213-140-4
UN: 2378
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=69c5150e0822157987c6925d8ddbbddfbb4bebe7&vid=69c5150e0822157987c6925d8ddbbddfbb4bebe7&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 96.0 %
Purity (Nonaqueous Titration) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR