Stampa…

5-(Tributylstannyl)pyrimidine 96%

Fornitore: Thermo Scientific
Pericolo

H51458.MD H51458.03
H51458.MDEA 72.4 EUR
H51458.MD H51458.03
5-(Tributylstannyl)pyrimidine 96%
5-(Tributylstannyl)pyrimidine

Purity: 96% CAS No.: 144173-85-3 Molecular Formula: C16H30N2Sn Formula Weight: 369.13 UN No.: 2788 MDL No.: MFCD01319056

Formula: C₁₆H₃₀N₂Sn
Peso molecolare: 369,13 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD01319056
CAS Number: 144173-85-3
ADR: 6.1,III

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR