(±)-Isoborneol ≥90.0% (by GC)
Fornitore: TCI
|
Attenzione
|
I0275-25G
I0275-500G
TCIAI0275-25GEA
16
EUR
TCIAI0275-25G
TCIAI0275-500G
(±)-Isoborneol ≥90.0% (by GC)
(±)-Isoborneol
Formula:
C₁₀H₁₈O |
MDL Number:
MFCD00074821 CAS Number: 124-76-5 EINECS: 204-712-4 Merck Index: 14,05128 |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 90.0 % |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...