Stampa…

3-Amino-N-methylbenzamide ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3-Aminobenzoylmethylamide

A1067-25G A1067-5G
TCIAA1067-25GEA 152 EUR
TCIAA1067-25G TCIAA1067-5G
3-Amino-N-methylbenzamide ≥98.0% (by HPLC, total nitrogen)
3-Amino-N-methylbenzamide
Formula: C₈H₁₀N₂O
Peso molecolare: 150,18 g/mol
MDL Number: MFCD00070630
CAS Number: 25900-61-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=54bd0b8f040077fa6beb17758b21ab933d4288cc&vid=54bd0b8f040077fa6beb17758b21ab933d4288cc&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light red powder to crystaline
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 118.0 to 121.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR