Stampa…

3-Decylthiophene ≥98.0% (by GC)

Fornitore: TCI EUROPE

D2098-1G D2098-5G
TCIAD2098-1GEA 45 EUR
TCIAD2098-1G TCIAD2098-5G
3-Decylthiophene ≥98.0% (by GC)
3-Decylthiophene
Formula: C₁₄H₂₄S
Peso molecolare: 224,40 g/mol
MDL Number: MFCD00143180
CAS Number: 65016-55-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b189a36c4a8c1789c572117a2c6257900fc24406&vid=b189a36c4a8c1789c572117a2c6257900fc24406&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR