Stampa…

Methyl-2-aminoisonicotinate ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Methyl 2-aminopyridine-4-carboxylate, Methyl 2-aminoisonicotinate

M2172-1G M2172-5G
TCIAM2172-1GEA 25 EUR
TCIAM2172-1G TCIAM2172-5G
Methyl-2-aminoisonicotinate ≥98.0% (by GC, titration analysis)
Methyl-2-aminoisonicotinate
Formula: C₇H₈N₂O₂
Peso molecolare: 152,15 g/mol
MDL Number: MFCD04039316
CAS Number: 6937-03-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=18a19e54d5723d1a4c3298dcc695bd47091306da&vid=18a19e54d5723d1a4c3298dcc695bd47091306da&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 145.0 to 149.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR