1-Bromo-4-dodecylbenzene ≥95.0% (by GC)
Fornitore: TCI EUROPE
B1802-25G
B1802-5G
TCIAB1802-25GEA
501
EUR
TCIAB1802-25G
TCIAB1802-5G
1-Bromo-4-dodecylbenzene ≥95.0% (by GC)
1-Bromo-4-dodecylbenzene
Formula:
C₁₈H₂₉Br Peso molecolare: 325,33 g/mol Punto di ebollizione: 191°C |
MDL Number:
MFCD00191386 CAS Number: 126930-72-1 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...