Stampa…

7-Tetradecanol ≥96.0% (by GC)

Fornitore: TCI EUROPE

T1759-1G T1759-5G
TCIAT1759-1GEA 88 EUR
TCIAT1759-1G TCIAT1759-5G
7-Tetradecanol ≥96.0% (by GC)
7-Tetradecanol
Formula: C₁₄H₃₀O
Punto di ebollizione: 156°C
MDL Number: MFCD00049210
CAS Number: 3981-79-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=27e5bde39c645b8c146578741a738afb4d2f4cc6&vid=27e5bde39c645b8c146578741a738afb4d2f4cc6&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 96.0 %
Melting point 41.0 to 44.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR