Stampa…

5-Bromo-2-hydroxybenzonitrile ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

B2214-25G B2214-5G
TCIAB2214-25GEA 571 EUR
TCIAB2214-25G TCIAB2214-5G
5-Bromo-2-hydroxybenzonitrile ≥98.0% (by GC, titration analysis)
5-Bromo-2-hydroxybenzonitrile
Formula: C₇H₄BrNO
Peso molecolare: 198,02 g/mol
MDL Number: MFCD00143428
CAS Number: 40530-18-5
EINECS: 254-958-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=089050d700997597c7d73c31c3a5174952774f55&vid=089050d700997597c7d73c31c3a5174952774f55&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 160.0 to 164.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR