Ethyl trifluoropyruvate ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Ethyl 3,3,3-trifluoro-2-oxopropanoate, Ethyl 3,3,3-trifluoropyruvate
T1434-25G
T1434-5G
TCIAT1434-25GEA
48
EUR
TCIAT1434-25G
TCIAT1434-5G
Ethyl trifluoropyruvate ≥98.0% (by GC)
Ethyl trifluoropyruvate
Formula:
C₅H₅F₃O₃ Peso molecolare: 170,09 g/mol Flash Pt: 31°C |
MDL Number:
MFCD00114935 CAS Number: 13081-18-0 UN: 1993 ADR: 3,III |
Risultati del test di specificazione
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...