Stampa…

2-Methoxyisonicotinic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

M2079-1G M2079-5G
TCIAM2079-1GEA 30 EUR
TCIAM2079-1G TCIAM2079-5G
2-Methoxyisonicotinic acid ≥98.0% (by GC, titration analysis)
2-Methoxyisonicotinic acid
Formula: C₇H₇NO₃
Punto di fusione: 203°C
MDL Number: MFCD04115718
CAS Number: 105596-63-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1bd381cd375da4e5bb1949f691bd615a38c90f45&vid=1bd381cd375da4e5bb1949f691bd615a38c90f45&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR