5-Methylbenzoxazole ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
5-Methylbenzo[d]oxazole
M1688-25G
M1688-5G
TCIAM1688-25GEA
590
EUR
TCIAM1688-25G
TCIAM1688-5G
5-Methylbenzoxazole ≥98.0% (by GC)
5-Methylbenzoxazole
Formula:
C₈H₇NO Peso molecolare: 133,15 g/mol Punto di ebollizione: 96°C |
MDL Number:
MFCD00216938 CAS Number: 10531-78-9 |
Risultati del test di specificazione
Appearance | White to Green to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 43.0 to 47.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...