4-(Chloromethyl)benzonitrile ≥97.0% (by GC, titration analysis)
Fornitore: TCI
|
Pericolo
|
Formula:
C₈H₆ClN Punto di ebollizione: 263°C |
MDL Number:
MFCD00019761 CAS Number: 874-86-2 EINECS: 212-869-5 UN: 2923 ADR: 8,II |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Argentmetric Titration) | min. 97.0 % |
Melting point | 78.0 to 82.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...