Stampa…

Ethyl dibromoacetate 96%

Fornitore: Thermo Fisher Scientific
Pericolo

B23209.06 B23209.14
B23209.06EA 84 EUR
B23209.06 B23209.14
Ethyl dibromoacetate 96%
Ethyl dibromoacetate

Purity: 96% CAS No.: 617-33-4 Molecular Formula: C4H6Br2O2 Formula Weight: 245.90 Refractive Index: 1.4990 Boiling Point: 76-78°/12mm MDL No.: MFCD00041718

Formula: C₄H₆Br₂O₂
Punto di ebollizione: 76…78 °C (12 mmHg)
Densità: 1,902
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00041718
CAS Number: 617-33-4
EINECS: 210-510-7

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR