Stampa…

2-Benzyloxy-5-fluorophenylboronic acid (contains varying amounts of Anhydride)

Fornitore: TCI
Attenzione

Synonyms: (2-(Benzyloxy)-5-fluorophenyl)boronic acid, 2-Benzyloxy-5-fluorobenzeneboronic acid

B3683-1G B3683-5G
TCIAB3683-1GEA 28 EUR
TCIAB3683-1G TCIAB3683-5G
2-Benzyloxy-5-fluorophenylboronic acid (contains varying amounts of Anhydride)
2-Benzyloxy-5-fluorophenylboronic acid
Formula: C₁₃H₁₂BFO₃
MDL Number: MFCD03788403
CAS Number: 779331-47-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2570a4217f7a741e5f3e51931bfb0ec4b6a0499f&vid=2570a4217f7a741e5f3e51931bfb0ec4b6a0499f&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 107.9 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR