Stampa…

3-(Benzyloxy)phenylboronic acid (contains varying amounts of Anhydride)

Fornitore: TCI EUROPE
Attenzione

Synonyms: (3-(Benzyloxy)phenyl)boronic acid, 3-Benzyloxybenzeneboronic acid

B3056-25G B3056-5G
TCIAB3056-25GEA 227 EUR
TCIAB3056-25G TCIAB3056-5G
3-(Benzyloxy)phenylboronic acid (contains varying amounts of Anhydride)
3-(Benzyloxy)phenylboronic acid
Formula: C₁₃H₁₃BO₃
Peso molecolare: 228,06 g/mol
MDL Number: MFCD02093063
CAS Number: 156682-54-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=12b6cc07da148fd0ba2c888f9eefe38b8da2b882&vid=12b6cc07da148fd0ba2c888f9eefe38b8da2b882&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Orange powder to crystal
Purity (Neutralization titration) 97.0 to 109.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR