Stampa…

2-Iodosobenzoic acid ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

I0073-10G I0073-1G
TCIAI0073-10GEA 551 EUR
TCIAI0073-10G TCIAI0073-1G
2-Iodosobenzoic acid ≥98.0% (by titrimetric analysis)
2-Iodosobenzoic acid
Formula: C₇H₅IO₃
Peso molecolare: 264,02 g/mol
MDL Number: MFCD00002401
CAS Number: 304-91-6
EINECS: 206-159-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eaa2302e692287286c912a41004605124f2d18f5&vid=eaa2302e692287286c912a41004605124f2d18f5&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Iodometric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR